To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1B
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:DYRK1B
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:DYRKB|Mirk protein kinase|AOMS3|MIRK|Dual-specificity tyrosine-phosphorylation regulated kinase 1B
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10085-596
- Supplier No.:12240-1-AP
Specifications
About this item
DYRK1B(dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1B) is also named as MIRK. It is a member of the dyrk/minibrain family of serine/threonine kinases that mediate the transition from growth to differentiation in lower eukaryotes and mammals. DYRK1B functions as a transcriptional coactivator that is activated by co-expressed MKK3, a MAP kinase that also can activate p38 MAP kinase. The DYRK1B splice variants found in each muscle tissue exhibit approximate molecular weights of 70 and 75 kDa. It has 3 isoforms produced by alternative splicing of the molecular weight of 65-69 kDa.
Western Blot: Mouse Testis Tissue, 1:1000-1:10000
Type: Primary
Antigen: DYRK1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat