To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:FXYD Domain Containing ion transport regulator 5
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:FXYD5
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:DYSAD|FXYD domain-containing ion transport regulator 5|IWU1|HSPC113|OIT2|keratinocytes associated transmembrane protein 1|RIC|IWU-1|dysad|KCT1|Dysadherin|PRO6241
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10085-600
- Supplier No.:12166-1-AP
Specifications
About this item
Dysadherin, also known as FXYD5 (FXYD domain containing ion transport regulator 5), is a cancer-associated cell membrane glycoprotein which belongs to the FXYD family. The FXYD family, which contains seven members, are tissue specific regulators of the Na,K-ATPase. Dysadherin is involved in down-regulation of E-cadherin, which plays important roles in tumor development and metastasis. It is present in spleen, lung, skeletal muscle, and testis. Increased expression of dysadherin has been associated with increased cell motility and metastatic potential.
Western Blot: Mouse small intestine Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200
Type: Primary
Antigen: Dysadherin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse