To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:NCK-Associated Protein 1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:NCKAP1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10090-870
- Supplier No.:12140-1-AP
Specifications
About this item
NCK-associated protein 1 (NCKAP1), also known as NAP1 or HEM2, is encoded by an apoptosis-related gene human NAP1, the expression of which is strongly down-regulated in sporadic Alzheimer's disease (AD). Human NAP1 is predominantly expressed in neuronal cells. Antisense oligo DNA of human NAP1 transcripts was found to induce apoptosis of neuronal cells. Loss of NAP1 function disrupts neuronal differentiation. Likewise, NAP1 plays an essential role in facilitating neuronal cytoskeletal changes underlying the postmigratory differentiation of cortical neurons, a critical step in functional wiring of the cortex. Human NAP1 is also proved to be an orthologue of rat Nap1 which binds to the adaptor molecule NCK in signal transduction.
Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human gliomas Tissue, 1:20-1:200
Type: Primary
Antigen: NCKAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat