To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Pre-B-cell leukemia Homeobox interacting Protein 1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:PBXIP1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10091-896
- Supplier No.:12102-1-AP
Specifications
About this item
PBXIP1 (pre-B-cell leukemia transcription factor-interacting protein 1), also known as HPIP (hematopoietic PBX-interacting protein), is a regulator of pre-B-cell leukemia transcription factors (BPXs) function. PBXIP1 is a nucleo-cytoplasmic shuttling protein, mainly localized in the cytosol and in small amounts in the nucleus . It interacts with PBX1 as well as PBX2 and PBX3. PBXIP1 inhibits the ability of PBX-HOX heterodimers to bind to target sequences and strongly inhibits the transactivation activity of E2A-PBX. PBXIP1 is also reported to be a microtubule-binding protein, which regulates estrogen receptor functions and plays a role in cancer development and progression .
Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; IP: HeLa Cells, 1:200-1:2000
Type: Primary
Antigen: PBXIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human