To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Von Willebrand Factor
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:VWF
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:F8VWF|Von Willebrand antigen II|vWF|VWD
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10097-058
- Supplier No.:11778-1-AP
Specifications
About this item
Von Willebrand factor (vWF) is a large multimeric glycoprotein that is crucial to the hemostasis process. It supports platelet adhesion and carries factor VIII (FVIII). Deficiency of vWF results in von Willebrand disease (vWD), a common inherited bleeding disorder. The pre-pro-vWF is comprised of 2,813 amino acids (aa) which encompass a 22-aa signal peptide, a 741-aa large propeptide (vWF propeptide, vWFpp, also called von Willebrand antigen II) and 2,050 aa making up the mature vWF. The von Willebrand antigen 2 is required for multimerization of vWF and for its targeting to storage granules. This antibody raised against the N-terminal region (residues 56-256 aa) of pre-pro-vWF recognizes von Willebrand antigen 2 (75-83 kDa) and pro-vWF (309-320 kDa).
Western Blot:Jurkat Cells, 1:200-1:2000; IHC: Human Liver Tissue, 1:20-1:200
Type: Primary
Antigen: VWF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat