To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Intelectin 1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ITLN1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:hIntL|Intestinal Lactoferrin Receptor|HL-1|LFR|omentin|ITLN|INTL|Galactofuranose-binding Lectin|Intelectin-1|HL1|Endothelial Lectin HL-1
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10089-038
- Supplier No.:11770-1-AP
Specifications
About this item
ITLN1, also named as INTL, ITLN and LFR, has no effect on basal glucose uptake but enhances insulin-stimulated glucose uptake in adipocytes. It may play a role in the defense system against microorganisms. ITLN1 may be involved in iron metabolism. The expression levels of ITLN1 is significantly different between the GPG and PPG (p<0.05).
Western Blot: Mouse Colon Tissue, 1:500-1:5000; IHC: Human small intestine Tissue, 1:20-1:200
Type: Primary
Antigen: ITLN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat