To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- ChIP:Yes
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:HBP1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10088-080
- Supplier No.:11746-1-AP
Specifications
About this item
HMG-box protein 1 (HBP1) is a transcriptional repressor, and has a role in cell cycle progression and tumor suppression. Also it's a repressor of the cyclin D1 gene and inhibits the Wnt signaling pathway. Interaction with RB1 enhances it's binding to the H1F0 promoter . Disrupts the interaction between DNA and TCF4. This is a rabbit polyclonal antibody raised against part of C-terminus of HBP1 of human origin.
Western Blot: Human Brain Tissue, 1:500-1:5000
Type: Primary
Antigen: HBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human