To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:High-mobility Group Nucleosome Binding Domain 1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:HMGN1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:MGC117425|FLJ31471|MGC104230|HMG14|FLJ27265
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10088-342
- Supplier No.:11695-1-AP
Specifications
About this item
HMGN1 (high-mobility group nucleosome-binding protein 1) is a member of the HMG superfamily of nonhistone chromatin-binding proteins, which which regulate chromatin structure and gene transcription. It binds to the inner side of the nucleosomal DNA thus altering the interaction between the DNA and the histone octamer. It may participate in the process which maintains transcribable genes in an unique chromatin conformation, and inhibit the phosphorylation of nucleosomal histones H3 and H2A by RPS6KA5/MSK1 and RPS6KA3/RSK2 expressed abundantly
Western Blot: HepG2 Cells, 1:500-1:5000; IF: HepG2, 1:10-1:100
Type: Primary
Antigen: HMGN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat