To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin carboxyl-terminal hydrolase isozyme L5
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:UCHL5
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:(Ubiquitin C-terminal hydrolase L5|Ubiquitin C-terminal hydrolase|CGI-70|UCH-L5|Ubiquitin thioesterase L5|AD-019|UCH37
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10096-708
- Supplier No.:11527-1-AP
Specifications
About this item
UCHL5(Ubiquitin carboxyl-terminal hydrolase isozyme L5) is also named as UCH37 and belongs to the peptidase C12 family. It isfunctionally linked with multiple protein complexes and signal transduction pathways. Uch37’s weight average molecular weight increases as a function of its concentration. By size-exclusion chromatography, the apparent molecular weight of Uch37 was 51, 56, 70, and 109 kDa at 0.15, 0.69, 2.4, and 9.5 mg/mL, respectively. Given Uch37’s monomeric molecular weight of 37 kDa and its apparent elongated shape, its tendency toward an apparent molecular weight between 40 and 50 kDa at decreasing concentrations is appropriate for Uch37 monomers.
IHC: Human Colon cancer Tissue, 1:20-1:200; Western Blot: Mouse Colon Tissue, 1:200-1:1000
Type: Primary
Antigen: UCHL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat