To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Myozenin 2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:MYOZ2
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:C4orf5|CMH16|CS-1
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10090-776
- Supplier No.:11450-1-AP
Specifications
About this item
MYOZ2 gene encodes for the calcineurin-binding protein myozenin-2 (or Calsarcin-1) belonging to myozenin family. Defects in MYOZ2 gene are the cause of familial hypertrophic cardiomyopathy type 16 (CMH16), a hereditary heart disorder characterized by ventricular hypertrophy. The disorder has inter- and intrafamilial variability ranging from benign to malignant forms with high risk of cardiac failure and sudden cardiac death . Myozenins may serve as intracellular binding proteins involved in linking Z line proteins such as alpha-actinin, and gamma-filamin. Important role in the modulation of calcineurin signaling and myofibrillogenesis of myozenin-2 have been elucidated . Proteintech’s Myozenin-2 antibody 11450-1-AP was raised against full-length protein of human origin and is able to detect band of approximate 34 kDa in Western blotting.
Western Blot: Human Brain Tissue, 1:500-1:5000
Type: Primary
Antigen: MYOZ2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat