To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome c oxidase subunit 5a
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:COX5A
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10085-060
- Supplier No.:11448-1-AP
Specifications
About this item
COX5A(Cytochrome c oxidase subunit 5A,mitochondrial)is also named as COXVA and belongs to the cytochrome c oxidase subunit 5A family.This is the heme A-containing chain of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.The mature human COX Va polypeptide shares 35% and 95% protein sequence identity with the corresponding S. cerevisiae COX subunit VI and bovine COX Va, respectively.
IHC: Human gliomas Tissue, 1:20-1:200; Western Blot: HeLa Cells, 1:200-1:2000
Type: Primary
Antigen: COX5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: