To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Rabphilin 3A
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:RPH3A
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoFluorescence:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:Rabphilin 3A homolog mouse
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10093-896
- Supplier No.:11396-1-AP
Specifications
About this item
RAB3A is a small G protein that is thought to act in exocytosis of neurotransmitters and hormones in synaptic neurotransmission and cell-cell communication. RPH3A (RPH3A), a RAB3A effector, is a neuronal C2 domain tandem containing protein that is involved in the neuronal transmitter process. Huntington's disease (HD) is a hereditary neurodegenerative disorder characterized by progressive motor disturbances, cognitive defects and dementia. Level of rabphilin 3A is substantially decreased in synapses of most brain regions in R6/1 transgenic mouse model of HD suggesting a role of RPH3A in impaired synaptic transmission.
Western Blot:COLO 320 Cells, 1:200-1:2000; IF:COLO 320 Cells, 1:10-1:100
Type: Primary
Antigen: RPH3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat