To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:DEXH (Asp-Glu-X-His) box Polypeptide 58
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:LGP2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:DEXH (Asp-Glu-X-His) box polypeptide 58 |RNA helicase LGP2 |D11LGP2 |DHX58|D11lgp2e |LGP2|ortholog of mouse D11lgp2
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10085-846
- Supplier No.:11355-1-AP
Specifications
About this item
DHX58(Probable ATP-dependent RNA helicase DHX58) is also named as D11LGP2E, LGP2 and belongs to the RLR subfamily. DHX58, originally identified as a highly expressed gene in mammary tumors, is another cytoplasmic DEX(D/H)-box helicase that can recognize RNA. It acts as a positive, but not negative, regulator of RIG-I– and MDA5-dependent recognition of RNA virus infection and plays a pivotal role in antiviral responses in vivo.
Western Blot: HEK-293 Cells, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:10-1:100; IP: Mouse Liver Tissue, 1:200-1:1000
Type: Primary
Antigen: LGP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat