To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Xeroderma Pigmentosum, complementation group G
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:XPG
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:ERCM2|XPG|OTTHUMP00000064902|COFS3|UVDR|XPGC|ERCC5
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10097-178
- Supplier No.:11331-1-AP
Specifications
About this item
The human genes correcting DNA repair defects are termed excision-repair cross-complementing or ERCC genes. The ERCC5 gene corrects the excision repair deficiency of Chinese hamster ovary cell line UV135 of complementation group 5. The human ERCC5 gene product is a structure-specific endonuclease required for making the 3-prime incision during DNA nucleotide excision-repair (NER). It also plays an important role in regulating DNA excision repair, removal of bulky lesions caused by environmental chemicals or UV light .
Western Blot: HeLa Cells, 1:200-1:2000
Type: Primary
Antigen: XPG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human