To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Eyes Absent homolog 2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:EYA2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:EYA2|EAB1|EAB 1|Translation of this uORF probably lowers the translation efficiency of EYA2.|EYA 2|eyes absent 2 homolog Drosophila|Eyes absent homolog 2|EYA2_HUMAN
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10086-738
- Supplier No.:11314-1-AP
Specifications
About this item
EYA2, also named as EAB1, belongs to the HAD-like hydrolase superfamily and EYA family. It coactivates SIX1. The Six1-EYA2 interaction may inhibit breast cancer progression. EYA2 is required for Six1-activated TGF-beta signaling. It may be involved in development of the eye. This protein has 3 isoforms produced by alternative splicing with the molecular weight of 59 kDa, 57 kDa and 50 kDa. It can be detected two bands of 70 kDa and 56 kDa in the neuroblastoma cells by western blot.
Western Blot: HeLa Cells, 1:200-1:1000; IHC: Human Prostate Cancer Tissue, 1:20-1:200
Type: Primary
Antigen: EYA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat