To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:NIMA-related kinase 9
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:NEK9
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:NERCC1|Nek8|NERCC
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10091-020
- Supplier No.:11192-1-AP
Specifications
About this item
NEK9 is also named as KIAA1995, NEK8, NERCC and belongs to the NEK Ser/Thr protein kinase family. It is originally identified as αβ-casein kinase that associates with a putative substrate Bicd2. It is also independently identified as a Nek6- and Ran GTPase-binding protein under a different name, Nercc1. Nek9, together with the highly similar (80% identical) Nek6 and Nek7, form a signaling module that is activated during mitosis and involved in the regulation of the mitotic spindle. The endogenous Nek9 (with predicted molecular mass of 120 kDa) has an apparent molecular mass of 600 kDa, also compatible with a tetramer.
Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Lymphoma Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200; FC: HepG2 Cells, N/A
Type: Primary
Antigen: NEK9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat