To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Isopentenyl-Diphosphate Delta Isomerase 1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:IDI1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:IPPI1|IPP1
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10088-578
- Supplier No.:11166-2-AP
Specifications
About this item
IDI1(Isopentenyl-diphosphate Delta-isomerase 1) is a peroxisomally-localized enzyme that catalyzes the interconversion of isopentenyl diphosphate (IPP) to its highly electrophilic isomer, dimethylallyl diphosphate (DMAPP). It belongs to the IPP isomerase type 1 family. This protein has 2 isoforms produced by alternative splicing with the molecular weight of 26 kDa and 32 kDa.
Western Blot: Mouse Liver Tissue, 1:200-1:1000
Type: Primary
Antigen: IDI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat