To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cell Division Cycle 34
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:CDC34
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10084-476
- Supplier No.:10964-2-AP
Specifications
About this item
CDC34 is also named as UBCH3, UBE2R1(Ubiquitin-conjugating enzyme E2 R1) and belongs to the ubiquitin-conjugating enzyme family. Cdc34 enzyme is itself a substrate for the type of polyubiquitin modification that is known to cause targeting of proteins for degradation. It is phosphorylated in vivo, providing yet another potential means for control of its activity. And the predominantly nuclear localization of this enzyme suggests that substrate molecules relevant to its cell cycle function are similarly located within the nucleus. Human CDC34 could substitute efficiently for yeast CDC34. It can form a homodimer.
Western Blot: Human pancreas Tissue, 1:200-1:1000; IHC: Human Prostate Cancer Tissue, 1:20-1:200
Type: Primary
Antigen: CDC34
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: