To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 25 Member A12
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:SLC25A12
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:solute carrier family 25|ARALAR1|CMC1_HUMAN|Araceli hiperlarga|Mitochondrial aspartate glutamate carrier 1|Calcium-binding mitochondrial carrier protein Aralar1|Solute carrier family 25 member 12|member 12.|ARALAR|Calcium binding mitochondrial carrier superfamily member|AGC1|SLC25A12
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10094-740
- Supplier No.:10789-1-AP
Specifications
About this item
SLC25A13, also known as citrin, has been identified as a mitochondrial aspartate-glutamate carrier protein and may be implicated in metabolic compartmentation. It consists of 675 amino acid residues and harbors four EF-hands and six mitochondrial transmembranous (TM) spanners. Mutations in SLC25A13 gene cause adult-onset type II citrullinemia (CTLN2) and neonatal intrahepatic cholestasis (NICCD). The complete deletion or cleaved SLA25A13 proteins have been reported in patients with SLC25A13 mutation.
Western Blot: Raji Cells, 1:200-1:2000; IHC: Human Colon cancer Tissue, 1:20-1:200
Type: Primary
Antigen: SLC25A12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat