To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Carboxypeptidase B2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:CPB2
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Antigen synonyms:TAFI|CPU|PCPB
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10085-102
- Supplier No.:10672-1-AP
Specifications
About this item
CPB2(Carboxypeptidase B2) belongs to the peptidase M14 family. The CPB2 gene encoding TAFI is a candidate gene for blood pressure. Circulating CPB2 and platelets may potentially contribute to regulating the bioactivity of leukocyte chemoattractant chemerin (RARRES2), and further extend the molecular link between blood coagulation/fibrinolysis and CMKLR1-mediated immune responses. This protein has 2 isoforms produced by alternative splicing.
Type: Primary
Antigen: CPB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: