To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Small Nuclear Ribonucleoprotein D3 Polypeptide 18kDa
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:SNRPD3
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10094-952
- Supplier No.:10379-1-AP
Specifications
About this item
Small nuclear ribonucleoprotein D3 polypeptide 18kDa (SNRPD3,synonym: SMD) belongs to the small nuclear ribonucleoprotein core protein family, with molecular weights of the D members: D1,16 kDa; D2, 16.5 kDa and D3 18 kDa. SNRPD3 is required for pre-mRNA splicing and small nuclear ribonucleoprotein biogenesis.
Western Blot: MCF7 Cells, 1:200-1:1000; IHC: Human small intestine Tissue, 1:20-1:200
Type: Primary
Antigen: SNRPD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat