To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:RAP1 GTPase-GDP Dissociation Stimulator 1
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:RAP1GDS1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:GDS1|SmgGDS
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10093-320
- Supplier No.:10377-1-AP
Specifications
About this item
GDS stimulates the GDP/GTP exchange reaction of a group of small GTP-binding proteins (G proteins), including at least c-Ki-ras p21, smg p21A, smg p21B, rhoA p21 and rhoB p21, by stimulating the dissociation of GDP from and the subsequent binding of GTP to each small G protein. The small GTPase Rap1, which is activated by a large variety of stimuli, functions in the control of integrin-mediated cell adhesion. RAP1GDS1 (synonym: GDS1)is one of RAP1 GTP-GDP dissociation stimulators.
Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human gliomas, 1:20-1:200
Type: Primary
Antigen: RAP1GDS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse