To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cholinergic receptor, nicotinic, alpha 3 (neuronal)
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:CHRNA3
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:PAOD2|NACHRA3|LNCR2
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10084-740
- Supplier No.:10333-1-AP
Specifications
About this item
CHRNA3, also named as NACHRA3, LNCR2 and PAOD2, is neuronal type nAChR subunits. It is a member of the nicotinic acetylcholine receptor family, which plays an important role in calcium regulation, neuronal development, and cognitive functions. Polymorphisms in this gene have been associated with an increased risk of smoking initiation and an increased susceptibility to lung cancer.
Western Blot: Mouse Thymus Tissue, 1:200-1:2000; IHC: Human Prostate Cancer Tissue, 1:20-1:200
Type: Primary
Antigen: CHRNA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: