Axygen® Robotic Tips for Corbett X-tractor, Corning
Catalog #: 89125-000
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Volume:200 µl
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Tip Color:Transparent
- Packed:96 Tips/Rack
- Filter:+
- Sterile:+
- Tip Style:Filtered Automation Tips
- For Use With:Corbett X-tractor Gene 8-Channel Head
- Supplier No.:CTF-199-R-S
Specifications
About this item
Tips in 96-well format designed for Corbett X-tractor Gene 8 channel head.
- Clear polypropylene
- With aerosol-resistant filters
- Optimised fine-point for accurate dispensing
- Certified RNase, DNase and endotoxin-free
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
Customers Who Bought This Also Bought