Arctic Express™ Dual Storage Systems, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Protect nonhazardous samples during shipment and storage using the Arctic Express storage systems
These dual shippers and storage systems feature excellent flexibility and safety features. Systems can be used as dry shippers as well as cryogenic stroage vessels.
Specifications
- Catalog No:
- 82005-416
- 82005-418
- 82005-420
- Model:
- Artic Express™ Dual 10
- Artic Express™ Dual 19
- Artic Express™ Dual 28
- Capacity:
- 10 L
- 18.5 L
- 28 L
- Ø neck:
- 5.08 cm (2")
- 5.6 cm (2.2")
- 9.7 cm (3.8")
- Ext.Ø:
- 25.9 cm (10.2")
- 36.8 cm (14.5")
- 46.2 cm (18.2")
Specifications
Customers Who Bought This Also Bought