OneRack® Tube Racks, Half Size
Supplier: Heathrow Scientific
Certificates
About this item
These durable tube racks are molded in a single, continuous piece so no assembly is required and there are no detachable pieces.
- Made out of fiberglass-reinforced polyoxymethylene (POM)
- Have molded coordinates for tube orientation
- Stable design for use in water baths
- Flat labeling area for easier identification
- Unique teardrop-shaped openings for better retention of tubes
- Offers the highest chemical and heat resistance (-40 to 121°C)
All racks feature molded coordinates to facilitate tube orientation and selection. Racks are autoclavable.
Tube size: 10 - 18 ml.
Customers Who Bought This Also Bought