To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Clonality:Polyclonal
- Gel shift:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:IgG F(c)
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:1.0 mg
- Western blot:Yes
- Epitope:F(c)
- Cross adsorption:No
- Format:Purified IgG
- Form:Clear, colorless liquid
- Antibody fragment:F(c)
- Storage buffer:30 mM Triethanolamine, pH 7.2, 5 mM Magnesium Chloride, 0.1 mM Zinc Chloride, 1 % (w/v) BSA, Protease/IgG free
- Storage temperature:2-8 °C
- Concentration:1.50 mg/ml (E 1% at 280 nm = 13.0)
- Shipping temperature:2-8 °C
- Immunogen:Mouse IgG Fc
- Purification:Affinity purified using solid phaseMouse IgG
- Cat. no.:10150-216
- Supplier No.:IR1903
Specifications
About this item
ImmunoReagents’ Alkaline Phosphatase (AP) antibody conjugates are used in ELISA, Immunohistochemistry (IHC) and Western blotting techniques. AP is a 140 kDa enzyme that changes color in the presence of PNPP or BCIP/NBT substrates. These conjugates have a linear reaction rate thus allowing the researcher to increase sensitivity by prolonging substrate incubation times.
- Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Mouse