To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Purified Chicken IgY Coupled to 4% Agarose Beads
- Size:2 mL
- Storage temperature:2 to 8°C, Do Not Freeze
- Cat. no.:10148-970
- Supplier No.:IR1008
Specifications
About this item
ImmunoReagents Chicken IgY Affinity Prep resins are prepared using highly purified species specific immunoglobulins
Immunoglobulins demonstrate a single arc when analyzed against normal serum proteins by IEP. The resins provided comprise of immunoglobulins covalently bound to a 4% agarose bead utilizing strong and stable hydrazide/aldehyde group chemistry. The Affinity Prep resins have 8-10 mgs of antibodies per/ml of resin, with a 24 month shelf life reagents.
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy. Antibodies produced cover a broad range of Immunoglobulin s from various species cross absorbed to provide high specificity.
This resin is suitable for removal of proteins recognizing Chicken IgY from liquid solutions, including traditional Immunoabsorptions and Immunoprecipitation protocols.