About this item
ImmunoReagents’ normal rabbit serum is used as a blocking agent or as a control for immunoassays
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy. Antibodies produced cover a broad range of Immunoglobulin s from various species cross absorbed to provide high specificity.
All serum is obtained from healthy, disease free animals. Serum is lipid extracted and buffer exchanged against phosphate buffer. Our serum is supplied in lyophilized form without preservative. It is recommended to reconstitute the normal serum with DI water and add a preservative of choice, or to aliquot reconstituted serum and store frozen to prevent contamination.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10152-238
- 10152-240
- Application:
- ELISA, WB, IHC, IP FACS, ID and Labeling
- ELISA, WB, IHC, IP FACS, ID and Labeling
- Storage temperature:
- Store lyophilized material at 2-8 °C. For long term storage after reconstitution, prepare small aliquots and store at -20 °C . For storage at 2-8 °C, add a preservative to prevent growth of bacteria.
- Store lyophilized material at 2-8 °C. For long term storage after reconstitution, prepare small aliquots and store at -20 °C . For storage at 2-8 °C, add a preservative to prevent growth of bacteria.