About this item
Designed for maximum product recovery for lab-scale or scale-up process volumes up to 4 L.
- Membrane Cassette Filter Media: Omega™ (low protein-binding, modified PES) membranes
- All product-wetted parts comply with pharmaceutical standards
- Easy connections through luer lock fitted ports with polished 316L stainless steel to ensure the same compatibility characteristics as production-scale Centrasette holders
Designed for maximum product recovery for lab-scale or scale-up process volumes up to 4 L.
Scale-up/scale-down device for development of tangential flow filtration (TFF) processes, biopharmaceutical small batch production, scale-down investigations, and production of materials for safety and efficacy studies (Phase I clinicals).
Identical path lengths provide linear scale-up to Pall Centramate and Centrasette™ cassette holders. The LV Centramate holder may be attached to other laboratory TFF systems to improve their scalability characteristics.
Low hold-up volume allows high concentration factors to be achieved from small starting volumes. A complete TFF system scalable to full production plant can be optimized using the volumes typically generated in the development or discovery lab.
All product-wetted parts comply with pharmaceutical standards
Easy connections through luer lock fitted ports with polished 316L stainless steel to ensure the same compatibility characteristics as production-scale Centrasette holders
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Related Information
Pall Laboratory Life Science Solutions
Product Literature
VWR GHS Compliance Letter (US)