About this item
Whatman Anotop 10 and 25 IC syringe filters from Cytiva's business are designed for efficient filtration prior to ion chromatography.
Inlet/outlet: Luer lock female/Luer male (except for Anotop™ 10 LC and Anotop™ 25 LC)
- Each batch is certified for low levels of anion leaching in ion chromatography (IC) filtration
- Filters large particulates, extending column life and increasing peak resolution
- Narrow pore size distribution and capillary pore structure for consistent results from a syringe filter
Whatman Anotop 10 and 25 IC syringe filters from Cytiva's business are designed for efficient filtration prior to ion chromatography.
Ion chromatography analysis often benefits from filtration as a sample preparation step. When done correctly, filtration can remove particles that might otherwise cause unwanted peaks or damage the chromatography column. Therefore, an ion exchange chromatography method should include a high-quality syringe filter.
Ion chromatography analysis often benefits from filtration as a sample preparation step. When done correctly, filtration can remove particles that might otherwise cause unwanted peaks or damage the chromatography column. Therefore, an ion exchange chromatography method should include a high-quality syringe filter.
Cytiva's Anotop IC syringe filters have an aluminum-based Anopore membrane that minimizes anion leaching during ion chromatography filtration. Leaching is validated at 10 to 30 ppb for major anions such as fluoride, chloride, bromide, sulfate, nitrite, and nitrate.
Anotop syringe filters are a versatile solution for most organic solvents and aqueous samples. Aluminum-based Anopore membranes are inorganic and designed to be inert and function in wide temperature ranges.
Anotop™ 10 IC:
Suitable for ion chromatography sample preparation and HPLC sample preparation.
Anotop™ 25 IC:
Suitable for ion chromatography sample preparation and HPLC sample preparation.
Specifications
- Catalog No:
- CA28297-720
- CA11013-912
- CA28138-053
- CA28138-055
- CA11013-020
- Type:
- Anotop™ 25 IC Syringe Filter
- Anotop™ 10 IC Syringe Filter
- Anotop™ 10 IC Syringe Filter
- Anotop™ 10 IC Syringe Filter
- Anotop™ 10 IC Syringe Filter
- Dimensions:
- 36.8 x 36.8 mm
- 15.4 x 18.5 mm
- 15.4 x 18.5 mm
- 15.4 x 18.5 mm
- 15.4 x 18.5 mm
- Hold-up volume:
- < 20 µl
- < 20 µl
- < 20 µl
- < 20 µl
- < 20 µl
- Max. operating pressure:
- 6.9 bar (100 psi)
- 6.9 bar (100 psi)
- 6.9 bar (100 psi)
- 6.9 bar (100 psi)
- 6.9 bar (100 psi)
Specifications
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
GE Healthcare Environmental testing for water air and soil Brochure (CAN) GE Healthcare Environmental testing for water air and soil Brochure (US) GE Healthcare Environmental Testing for Water Brochure GE Healthcare Environmental Testing for Water Brochure (CA) GE_Healthcare_Whatman_Filtration_Products_for_Pharmaceutical_Quality_Control_(US)