Whatman™ Weighing Papers, Grade 2122, Whatman products (Cytiva)
Supplier: Cytiva
Certificates
About this item
These Grade 2122 weigh paper sheets reliably allow samples to be transferred to scales without adding unwanted substances that could impact analytical results.
- Smooth surface allows samples to slide off with ease and not be retained on paper
- Shiny pergamyne paper allows visual determination that all sample has been removed
- Extremely thin (0.03 mm) and lightweight (40 g/m²) for safe and secure sample transport
- No glue or additives (residue-free)
This paper is not low in nitrogen. Therefore, it should not be used for Kjeldahl analysis.
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Customers Who Bought This Also Bought