3B Scientific® Deluxe Functional Knee Joint
Catalog #: 470103-140
Supplier: American 3B Scientific
Specifications
- Description:Deluxe Functional Knee Joint
- Cat. no.:470103-140
- Supplier No.:A82/1
- No. of Parts:1
Specifications
About this item
This high-quality deluxe functional knee joint model with ligaments shows the anatomy and possible physiological movements of the human knee joint in exceptional detail.
- Smaller size for easy storage
- Section of the joint on each base
- 3 year manufacturers warranty
- Includes 3B smart anatomy
This model clearly demonstrates abductions, anteversion, retroversion, internal and external rotation. This knee joint model also clearly defines the ACL and PCL.
The color of the natural-cast bones of the knee joint is extremely realistic. The cartilage on the knee joint surfaces is marked blue.
This model consists of portions of the femur, tibia and fibula, menisci and patella. The Functional Knee joint model is mounted on a base for easy display.
BodyTextAccessories: For information about 3B smart anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought