KIMBLE® Silicone Stoppers, DWK Life Sciences
Catalog #: KT774200-0022
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Color:Green
- Description:Blind hole stoppers
- Cat. no.:KT774200-0022
- Stopper No.:6
- Supplier No.:774200-0022
Specifications
About this item
Designed for use in centrifugal separators and gas sampling tubes. Blind hole stopper features easy syringe penetration.
Customers Who Bought This Also Bought