Whatman™ Grade GF/C Glass Microfiber Filters, Binder Free
Supplier: Cytiva
Certificates
About this item
These borosilicate glass filters are widely used as total suspended solids filters for applications testing both potable water and waste water. GF/C can also function as a filter for liquid scintillation counting, cell harvesting, industrial waste analysis and binding assays that require high loading capacity.
- Fine particle retention
- Good flow rate and loading capacity
- Binder free
Specifications
- Catalog No:
- CA28497-663
- CA28497-787
- CA28497-765
- CA28497-754
- CA28497-743
- CA28497-721
- CA28497-700
- CA11013-132
- CA11013-030
- CA11013-026
- CA14005-870
- CA11013-144
- CA28497-696
- CA11013-948
- CA28497-685
- CA28497-652
- CA28497-641
- CA11013-944
- CA28497-620
- CA28497-619
- CA28497-776
- Thickness:
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- 260 µm
- Particle retention:
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- 1.2 µm
- Filtration speed:
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Medium to fast, 100 s (Herzberg)
- Water flow rate:
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- 105 ml/min
- Airflow rate:
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- 6.7 s/100 ml/in²
- Basis weight:
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- 53 g/m²
- Max. operating temperature:
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
- 550 °C
Specifications
Related Information
Certificates
Search by Lot Number10035-308_16924209 28497-754_9767214 28497-765_17227340 28497-787_12857078 CA28497-696_17216723
Conformity declaration
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
GE Healthcare Environmental testing for water air and soil Brochure (US) GE Healthcare Environmental Testing for Water Brochure GE Healthcare Environmental Testing for Water Brochure (CA) GE Healthcare Whatman Laboratory Essentials Brochure GE_Healthcare_Whatman_Filtration_Products_for_Pharmaceutical_Quality_Control_(US)
Product Literature
Customers Who Bought This Also Bought