Sodium carbonate monohydrate 99.5-100.5% (by anhydrous basis), crystals NF, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Sodium carbonate hydrous
Formula:Na₂CO₃·1H₂O
MW:124.00 g/mol
Melting Pt:107.85 °C
Density:2.25 g/cm³ (20 °C)
MDL Number:MFCD00149177
CAS Number:5968-11-6
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 107.85 °C
:Flash Pt:
:Flash Pt:
:M.W. 124.00 g/mol
Storage Temperature:Ambient
EINECS:207-838-8
:GHS07
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GMP Manufactured Product:
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Meets N.F. Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Water (H₂O):12.0 - 15.0 %
- NF - Assay (Na₂CO₃) (anhydrous basis):99.5 - 100.5 %
- NF - Endotoxin Concentration (EU/g):≤ 2.5
- EP/BP - Assay (Na₂CO₃):83.0 - 87.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Alkali Hydroxides and Bicarbonates:Passes Test
- EP/BP - Chloride (Cl):≤ 125 ppm
- EP/BP - Sulfate (SO₄):≤ 250 ppm
- EP/BP - Iron (Fe):≤ 50 ppm
- No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
- J.T.Baker®, Sodium carbonate monohydrate 99.5-100.5% (by anhydrous basis), crystals NF, Multi-Compendial:
Specification Test Results
Related Information
Certificates
Search by Lot Number3603 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_JT3603-7
Product specification
Customers Who Bought This Also Bought