Connector Kit, Kimble Chase
Catalog #: 89004-516
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:Glass-filled Nylon Connectors with PTFE Seals
- Cat. no.:89004-516
- Supplier No.:747205-0000
Specifications
About this item
Connector kit with ten connectors; one each of the following sizes: 8-425 to 8-425; 8-425 to 13-425; 13-425 to 13-425; 13-425 to 15-425; 13-425 to 20-400; 15-425 to 20-400; 18-400 to 20-400; 20-400 to 20-400; 20-400 to 22-400; 20-400 to 24-400.
For use up to 150 °C. Connectors have PTFE seals, FKM o-rings and blue glass-filled nylon. The 24-400 cap is black polypropylene.
Customers Who Bought This Also Bought