Whatman™ Grade 598 Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Cytiva
New Product
Certificates
About this item
A thick filter paper with high loading capacity. Combines medium retention with medium-fast to fast filtration speed.
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Customers Who Bought This Also Bought