KIMBLE® Right Angle Flow Control Inlet Adapter with Glass Stopcock and Inner Standard Taper Joint, DWK Life Sciences
Catalog #: KT185000-2440
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- ST Joint:24/40
- Cat. no.:KT185000-2440
- Overall Height:130
- Fits Tubing I.D.:⅜"
- Supplier No.:185000-2440
Specifications
About this item
Use this adapter for regulating the input of gases or liquids into reaction systems.
- Stopcock with glass plug
- Inner [ST] joint on one end
- 90° angle hose connection on opposite end
Adapter features an inner Standard Taper joint on one end and a 90° hose connection on the opposite end
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Stopcock has a glass plug.
Customers Who Bought This Also Bought