About this item
Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapours.
Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapours. With integrated air valve for pressure equalisation. High quality materials (PTFE, PP) ensure maximum safety for your laboratory staff and environment.
- Especially recommended for HPLC Systems
- Chemically resistant against aggressive organic solvents
With integrated air valve for pressure equalisation. High quality materials (PTFE, PP) ensure maximum safety for your laboratory staff and environment.
SafetyCaps with shut-off
During repairs on HPLC pumps, the shut-off on the SafetyCaps can be closed to keep capillaries and inlet filters in the solvent. The frit does not come into contact with the ambient air. The pump can be quickly and easily flushed during purging after repairs. The SafetyCaps with shut-off also prevent air pockets in the tube when the supply bottle is being changed and guarantees that the analysis continues without a problem.
SafetyCaps for NS 29/32 ground neck bottles
The locknut allows the stopper to be easily loosened and removed, even after long usage. For bottles with NS 29/32 mm standard ground joints.
Related Information
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
SCAT Europe Safety Cap Test Report.pdf