ContactUsLinkComponent
KIMBLE® Plastic Luer Slip Micro Syringe, DWK Life Sciences
KIMBLE® Plastic Luer Slip Micro Syringe, DWK Life Sciences
Catalog #: KT748019-0001
Labsets kits Organic Chemistry Microscale Threaded / [standard taper] 14/10 Standard
KIMBLE® Plastic Luer Slip Micro Syringe, DWK Life Sciences
Catalog #: KT748019-0001
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Volume:
    1000 µl (1 ml)
  • Cat. no.:
    KT748019-0001
  • Graduations:
    0.01 cc
  • Supplier No.:
    748019-0001
Specifications
About this item
The Plastic Luer Slip Micro Syringes work for chromatography, nuclear medicine, HPLC sample preparation, and many laboratory procedures.
  • Two Part
  • Plastic
  • Polypropylene Barrel
  • Polyethylene Plunger
  • Latex Free
More chemically resistant than rubber-tipped syringes, these two-part plastic syringes feature a polypropylene barrel and polyethylene, low dead-space plunger
The needles are for research use only and are not for use in diagnostic procedures.
The sterile, individually wrapped needles contain no rubber, silicone oil, styrene, DNA, latex, or DEHP.
Ordering information: Syringes are supplied without needles.
Customers Who Bought This Also Bought