To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Ham’s F-12 without Hypoxanthine and L-Glutamine
- Size:1 L
- Sterility:Sterile filtered
- Shipping conditions:Ambient temperature
- Osmolality:295 ±5% mOsm/Kg H₂0
- Shelf Life:24 months
- Supplier No.:112-153-131
Specifications
About this item
Compared to other basal media, Ham’s F-12 contains a wider variety of components, including zinc, putrescine, hypoxanthine, and thymidine. Ham’s F-12 contains no proteins or growth factors, and uses a sodium bicarbonate buffer system (1.176 g/L); therefore requiring a 5–10% CO2 environment to maintain physiological pH.
- Without hypoxanthine and L-glutamine
Ham’s F-12 Nutrient Mixture without Hypoxanthine is useful for serum free clonal growth of Chinese Hamster Ovary (CHO) cells, chick embryos and mouse L-cells. Ham’s F-12 Nutrient Mixture without Hypoxanthine can also be used to culture myeloma cells and hybridomas, as well as primary rat hepatocytes and prostate epithelial cells.
Recommendation
Add L-glutamine prior to use at a final concentration of 1.0 mM.
Supplement 10% FBS because F-12 contains no proteins or growth factors.
Sterile Filtered
Harmonization Code: 3821.00.0000
ECCN: EAR99