To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Fischer's medium
- Size:500 ml
- Sterility:Sterile filtered
- Shipping conditions:Ambient temperature
- pH:7.0 ±0.2
- Shelf Life:24 months
- Supplier No.:112-032-101
Specifications
About this item
Fischer’s medium was originally developed by Glen Fischer at the National Cancer Institute (Bethesda, MD), for serial propagation of cells from leukemic mice. Fischer’s medium supports clonal growth of cells – particularly lymphoblasts – from primary explants or cells already in culture.
- With L-glutamine and Phenol Red
Fischer’s medium also supports mammalian cell culture and murine hematopoietic cells.
Recommendation
Supplement with 1.125 g/L of sodium bicarbonate for increased buffering capability and cell performance
Sterile Filtered
Harmonization Code: 3821.00.0000
ECCN: EAR99