KIMBLE® PTFE BEVEL-SEAL™ Inlet Adapter, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
This is an adjustable fitting designed to adapt to a variety of apparatus.
- Vacuum-tight seal
- Bottom [ST] inner joint
- Design allows use with clamps
- FKM O-Rings and polypropylene cap included
A diameter variation of 1mm to 2mm can be accommodated, as the inert O-Ring is compressed by the open-top cap to create a vacuum-tight seal. The adapter body is made of PTFE. Bottom has a Standard Taper inner joint. Design allows use with clamps. Supplied with FKM O-Rings and with polypropylene cap suitable for use to 150°C (302°F).
Customers Who Bought This Also Bought