Orders LinkContactUsLinkComponent
Plastic Bumper, KIMBLE®, DWK Life Sciences
Plastic Bumper, KIMBLE®, DWK Life Sciences
Bumper, size 4, fits 28.5 - 31.0 mm (100 ml) cylinder OD
Catalog # 89001-238
Supplier #:  20100-4
Plastic Bumper, KIMBLE®, DWK Life Sciences
Catalog # 89001-238
Plastic Bumper, KIMBLE®, DWK Life Sciences
Plastic Bumper, KIMBLE®, DWK Life Sciences
Bumper, size 4, fits 28.5 - 31.0 mm (100 ml) cylinder OD
Price
$26.60
New Customer Promotional Price
Log in for availability
Case of 6
Product Details & Documents
When placed around a cylinder near the top, this sturdy split ring protects against breakage if the cylinder is upset
Made of yellow low density polyethylene. Position of ring can be adjusted to avoid contact with the top of a receiver when liquid is poured from the cylinder. Should a ring become loose after prolonged use, soak it briefly in hot water (90 °C) to return it to its original shape.
Documents
Specifications
  • Description:
    Bumper, size 4, fits 28.5 - 31.0 mm (100 ml) cylinder OD