Transfer/Weighing Funnels, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Transfer / weighing funnels are ideal for easy transfer of liquids or powders
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Flat bottom allows weighing.
Customers Who Bought This Also Bought