KIMBLE® Threaded Compression Caps and Connectors, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Open-top compression cap and connectors consist of glass-filled nylon.
- Threaded
- For use to 200°C (392°F)
- PTFE seals
Connectors have PTFE seals.
Customers Who Bought This Also Bought