Specifications
- Description:VERSA 10 (8-channel) Liquid Handling Workstation
- Height:21"
- Connection:USB
- Cat. no.:CA11023-068
- Length:22"
- Weight:90 lbs.
- Width:18"
- Supplier No.:VER-10-8CH
Specifications
About this item
The VERSA 10 automated liquid handling workstation meets the needs of modern labs by providing the high throughput needed for many workflows with the compact size to fit into almost any lab space
Features include compact design to fit in almost any space, 6-position deck that allows versatile configuration, air displacement pipetting that gives the highest precision even at low volumes, 8-channel head to support high throughput applications, single channel functionality to support diverse sample numbers and applications and optional HEPA enclosure that protects samples from outside contamination.
The VERSA 10 Liquid Handling Workstation can be used in PCR and RT-PCR reaction setup, sequencing reaction setup, Nucleic Acid Purification, hit picking, serial dilution, enzymatic assays and general liquid handling applications.
The 8 channel liquid handling head features single channel functionality to allow the flexibility to perform a diverse array of applications on the same instrument. Liquid handling is carried out by a highly-accurate air displacement pipetting system to ensure the highest accuracy and reproducibility.