About this item
FUSION® successfully combines the flexibility, durability and appearance of Latex in a 100% latex allergen-free, dual-polymer stretch synthetic.
- The exterior glove layer is composed of FUSION elastomers, white pigment and high quality PVC
- The combination of these materials provides the flexible backbone of the FUSION glove
The interior surface is composed of polyurethane; widely recognized for its superior flexibility, durability, puncture resistance and non-allergenic properties. The successful combination of these materials results in the production of a glove with outstanding functional capabilities for medical, dental, laboratory or other demanding applications.
With the highest elasticity available in a stretch synthetic glove, FUSION provides the comfort and flexibility required for tasks that demand excellent tactile sensitivity, without restriction of hand movement or hand fatigue.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA32900-189
- CA32900-185
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Cuff style:
- Beaded
- Beaded
- Finger thickness:
- 5.0 mil
- 5.0 mil
- Palm thickness:
- 3.6 mil
- 3.6 mil