About this item
The HI100 antibody reacts with the human CD45 isoform known as CD45RA, a protein tyrosine phosphatase of 220 kDa. CD45 is one of the most abundant hematopoietic markers, and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). Various isoforms are generated and expressed in cell-specific patterns. With their broad cell distribution, CD45 isoforms are critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RB and CD45RO.
The HI100 antibody is widely used as a marker for human CD45RA expression on naïve and activated T cells, B cells, and monocytes.
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: HI100
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Chimpanzee
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10050-928
- 10050-930
- Antigen name:
- Protein Tyrosine Phosphatase Receptor Type C
- Protein Tyrosine Phosphatase Receptor Type C
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- HI100
- HI100
- Host:
- Mouse
- Mouse
- Gene ID:
- 5788
- 5788
- Isotype:
- IgG2b
- IgG2b